missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PD 4'-oxyacetic acid

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
PD 4'-oxyacetic is a carboxylic acid-functionalized cereblon ligand that can be used for PROTAC® Degrader development. The compound can be used to develop PD-PROTACs that exhibit improved stability and potency compared with PROTACs synthesized using IMiD (immunomodulatory imide drug) analogs.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 2-[4-(Tetrahydro-2,4-dioxo-1(2H)-pyrimidinyl)phenoxy]acetic acid |
| CAS | 2633633-12-0 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C12H12N2O5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction