Learn More
Tocris Bioscience™ PD 98059
GREENER_CHOICE
MEK inhibitor
Brand:  Tocris Bioscience™ 1213/10
Encompass
Product Code. 13498893
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.
Specifications
Specifications
98%
167869-21-8
10 mg
2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one
Room Temperature