missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PD 98059

Product Code. 18708126
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18708126 10 mg 10mg
18073852 1 mg 1mg
18033882 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18708126 Supplier R&D Systems™ Supplier No. 1213/10
  • ONLINE EXCLUSIVE PRICE
    264.99 €
    238.35 € / 10mg
    Save 26.64 €
    10% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MEK inhibitor

PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.
TRUSTED_SUSTAINABILITY

Specifications

CAS 167869-21-8
Quantity 10 mg
Product Line Tocris
Molecular Formula 2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one
Product Type PD 98059
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 267.28
Inhibitors PD 98059

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.