missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PD MidiTrap™ G-10

Product Code. 10075304 Shop All Cytiva Products
Click to view available options
Quantity:
Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10075304

Brand: Cytiva 28918011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleanup biological samples, such as peptides, small proteins, and oligosaccharides. Cytiva Life Sciences™ PD MidiTrap™ G-10 offers fast desalting and efficient removal of contaminants such as salts, dyes, and radioactive labels.

  • Prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides
  • Bed volume: 5.3mL
  • Bed dimensions: 13 x 40mm
  • Sample volume: 400 to 1000μL
Media:
  • Matrix: cross-linked dextran
  • Particle size wet: 55 to 165μm
  • Partical size dry: 40 to 120μm
  • pH stability working range: 2 to 13
Column:
  • Column I.D.: 13mm
  • Column material: polypropylene
  • Bed support material: polyethylene

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetate, pH 6, 0.1M NaOH, 70% Ethanol or 6M Guanidine Hydrochloride
Product Type PD MidiTrap G-10 gravity column
Bed Volume 5.3mL
Diameter (English) Inner 0.51 in.
Diameter (Metric) Inner 13mm
Quantity Each
Height (English) 1.5 in.
Height (Metric) 40mm
Flow Rate 24 to 49cm/hr.
Includes Instructions
Material Polyethylene
pH Range 2 to 13
Particle Size 40 to 120μm in dry form, 55 to 165μm in wet form
Support Type Cross-linked Dextran
Content And Storage 4° to 30°C
Column Type Gravity Column
For Use With (Application) Buffer exchange and cleanup of biological samples, for example Peptides, small Proteins and Oligosaccharides
Show More Show Less
compliance-icons
Signal Word
  • ||Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.