missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PD0325901
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
Description
- PD0325901 is a selective, cell permeable inhibitor of the MEK/ERK pathway that inhibits the activation and downstream signaling of MEK
- It is an extremely potent inhibitor, suppressing the phosphorylation of ERK in C26 cells at very low concentrations (IC50 = 0.33 nM) (Bain et al.), Barrett et al.)
Specifications
Specifications
| CAS | 391210-10-9 |
| Quantity | 25 mg |
| Molecular Formula | C16H14F3IN2O4 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction