missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies PD0325901

Product Code. 17100042
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
This item is not returnable. View return policy

Product Code. 17100042

Brand: STEMCELL Technologies 72184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MEK/ERK pathway inhibitor; Inhibits MEK

  • PD0325901 is a selective, cell permeable inhibitor of the MEK/ERK pathway that inhibits the activation and downstream signaling of MEK
  • It is an extremely potent inhibitor, suppressing the phosphorylation of ERK in C26 cells at very low concentrations (IC50 = 0.33 nM) (Bain et al.), Barrett et al.)

Specifications

CAS 391210-10-9
Quantity 10 mg
Molecular Formula C16H14F3IN2O4
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.