missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PDEdelta ATTEC 12C
Click to view available options
:
5 mg
Description
PDEδ ATTEC 12C is a selective PDEδ autophagy-tethering compound (ATTEC) Degrader (DC50 = 1.7 μM; Dmax = 85%; Kd = 56 nM). Degrades PDEδ through lysosome-mediated autophagy. Inhibits proliferation of KRAS mutant Capan-1 and MiaPaCa-2 pancreatic cancer cells (IC50 values are 0.8 μM and 1.4 μM respectively); dose- and time-dependently induces apoptosis.
Specifications
Specifications
| Chemical Name or Material | N-(6-(2-(2,6-Dibromo-4-((5-iodo-2-oxoindolin-3-ylidene)methyl)phenoxy)acetamido)hexyl)-4-(3,4-dimethyl-7-oxo-2-(p-tolyl)-2,7-dihydro-6H-pyrazolo[3,4-d]pyridazin-6-yl)butanamide |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C41H42Br2IN7O5 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction