Learn More
Fisherbrand™ PE Plug for Shell Vial
Product Code. 11997061 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
8 mm
12 mm
15 mm
Unit Size:
Pack of 100
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11997061 8 mm Pack of 1000
11804960 12 mm Pack of 1000
11556014 12 mm Pack of 100
11824980 15 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11997061
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Polyethylene caps are chemically inert and suitable for most chromatography applications.
  • PE Plug
  • Transparent
  • Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Diameter (Metric) Outer 8 mm
Material Polyethylene
Quantity 100
Closure Size ND8
Product Type Plug