missing translation for 'onlineSavingsMsg'
Learn More

Tork PeakServe™ Continuous Paper Hand Towels H5

Product Code. 30243719 Shop All Tork Products
Change view
Click to view available options
Color:
White
White, Gray
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243719 White Pack of 12
30243759 White, Gray Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30243719 Supplier Tork Supplier No. 100585
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 02-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Tork PeakServe™: the latest innovation designed for high-traffic washrooms where staff needs to maximize cleaning time and maintain a good flow of people.

  • Deliver high-quality performance and softness with our advanced quality Tork PeakServe™ Continuous hand towels.
  • Dry hands quickly - these paper hand towels are 20% more absorbent than Tork PeakServe™ universal quality refills.
  • Make a good impression on guests with the attractive leaf pattern of these paper hand towels.
  • Avoid delays in busy washrooms with our high-capacity dispenser system that ensures hand towels are always available.
TRUSTED_SUSTAINABILITY

Specifica

Product Type Hand Towel for Dispenser
Length (English) 7.91 in.
Length (Metric) 20.1 cm
Width (English) 8.86 in.
Width (Metric) 22.5 cm
Certifications/Compliance FSC Chain-Of-Custody, ISO 9001, OHSAS 18001, EU Ecolabel, EMAS, ISO 14001, FSC MIX Credit
Color White
Ply 1-ply
Certifications/Compliance (Europe) FSC Chain-Of-Custody, ISO 9001, OHSAS 18001, EU Ecolabel, EMAS, ISO 14001, FSC MIX Credit
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.