Learn More
Neogen Pecan Protein ELISA Kit, E96PEC
Description
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Specifications
| Product Type | Test |
| Quantity | 96 Wells |
| Test Time | 65 Minutes |