Learn More
Heinz Herenz Pediatric Tip Flocked VIR-Swab Virus Flocked Swab
Product Code. 30158434
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 600
Product Code. Sterility Unit Size
30158434 Sterile Pack of 600
1 options
Product Code. 30158434 Supplier Heinz Herenz Supplier No. 1020242
  • ONLINE EXCLUSIVE PRICE
    1369.86 €
    946.05 € / Pack of 600
    Save 423.81 €
    30% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Flocked swab with pediatric tip, test tube with red screw cap. Contains 1 mL of liquid transport medium developed for the collection, transportation and survival of viruses, chlamydia, mycoplasma and ureaplasma. Sterile.
Our transport and collection swabs contain special applicators and test tubes with different liquid media, customized for the transportation and preservation of microbiological samples.
  • The liquid medium allows the division of the sample material and thus several tests from one sample.
  • The ready to use devices are supplied in a peelpack which is easy to open.
  • The breakpoint in the shaft of the applicator enables a safe and hygienic workflow.
  • The standard size of the test tubes with screwcap simplified the use with automatic analyzers.
  • There are labels on the test tubes for a accurate sample identification.
  • The items are produced according to the conditions of GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485. This ensures a high quality.
  • Our swabs are sterile (SAL 106), pyrogen free and noncytotoxic.
Specifications
Product Type Universal Viral Transport
Format Swab
Media Type Neutralizers
Sterility Sterile
Color White