missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PEEK Connector 1/16 in. Luer

Product Code. 11370232 Shop All Cytiva Products
Change view
Click to view available options
Connections:
1/16 in. Tapered Male to Luer Female
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Unit Size
11370232 1/16 in. Tapered Male to Luer Female Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11370232 Supplier Cytiva Supplier No. 18111251
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Set of two (2) polyetheretherketone 1/16 in. Male/Luer Female connectors for connection of a syringe to the top of a chromatography column in the ÄKTA Chromatography System.

ÄKTA Laboratory-scale Systems are liquid chromatography systems used for protein purification at research laboratory scale.
  • Tapered syringe ends
  • Polyetheretherketone (PEEK) connstruction
  • Connections: 1/16 in. male–Luer female
TRUSTED_SUSTAINABILITY

Specifications

Type Connector
Connections 1/16 in. Tapered Male to Luer Female
Quantity Set of 2
Material PEEK
Color Black
For Use With (Equipment) ÄKTA Laboratory-Scale Systems, Syinge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.