missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 100 ft

Product Code. 15770999
Change view
Click to view available options
Diameter (English) Inner:
0.0025 in.
0.004 in.
0.005 in.
0.007 in.
0.010 in.
0.013 in.
0.015 in.
0.020 in.
0.030 in.
0.040 in.
Color:
Black
Blue
Gray
Green
Natural
Orange
Red
Yellow
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Color unitSize
15770999 0.004 in. Black Each
16642562 0.0025 in. Natural Each
15711009 0.005 in. Red Each
15751009 0.007 in. Yellow Each
15791009 0.010 in. Blue Each
15731019 0.013 in. Orange Each
15771019 0.015 in. Gray Each
16682562 0.015 in. Natural Each
15711029 0.020 in. Orange Each
15791029 0.030 in. Green Each
15751029 0.030 in. Red Each
15731039 0.040 in. Natural Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 15770999 Supplier Trajan Scientific and Medical Supplier No. 1302004006100F
Add to basket
Add to basket
This item is not returnable. View return policy

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Color Black
pH Range 2 to 14
Diameter (English) Inner 0.004 in.
Diameter (English) Outer 1/16 in.
Overall Length (English) 100 ft.
Material PEEK
Maximum Pressure 7000 psi (483 bar)
Max. Operating Temperature (Metric) 100°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.