missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 50 ft

Product Code. 16682552
Change view
Click to view available options
Diameter (English) Inner:
0.0025 in.
0.004 in.
0.005 in.
0.007 in.
0.010 in.
0.015 in.
0.020 in.
Color:
Black
Blue
Gray
Natural
Orange
Red
Yellow
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Color Unit Size
16682552 0.004 in. Black Each
16642552 0.0025 in. Natural Each
15790979 0.005 in. Red Each
15730989 0.007 in. Yellow Each
15770989 0.010 in. Blue Each
15710999 0.015 in. Gray Each
16622562 0.015 in. Natural Each
15750999 0.020 in. Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16682552 Supplier Trajan Scientific and Medical Supplier No. 130100400650F
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Color Black
Diameter (English) Inner 0.004 in.
Diameter (English) Outer 1/32 in.
Overall Length (English) 50 ft.
Maximum Pressure 5000 psi (345 bar)
Max. Operating Temperature (Metric) 100°C

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.