missing translation for 'onlineSavingsMsg'
Learn More

PEG4-azide, TRC

Product Code. 30335206
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335206 500 mg 500mg
30347052 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30335206 Supplier Toronto Research Chemicals Supplier No. TRCA885250500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Azido-3,6,9-trioxaundecane-11-ol
CAS 86770-67-4
Molecular Formula C8 H17 N3 O4
Quantity 500 mg
Synonym 2-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethoxy]ethanol
InChI Formula InChI=1S/C8H17N3O4/c9-11-10-1-3-13-5-7-15-8-6-14-4-2-12/h12H,1-8H2
SMILES OCCOCCOCCOCCN=[N+]=[N-]
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanol
Molecular Weight (g/mol) 219.24
Formula Weight 219.1219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.