missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress PEG6-(CH2CO2H)2

Product Code. 16495786
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495786 100 mg 100mg
16441287 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495786 Supplier MedChemExpress Supplier No. HY122702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PEG6-(CH2CO2H)2 is a symmetric PEG PROTAC linker, for the synthesis of Homo-PROTACs which is bivalent small-molecule dimerizers of the VHL E3 ubiquitin ligase to induce self-degradation.

Specifications

Molecular Weight (g/mol) 354.35 g/mol
CAS 77855-76-6
Content And Storage 4°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Shipping Condition Room temperature
Quantity 100 mg
Solubility Ethanol : 100 mg/mL (282.21 mM; Need ultrasonic)
Target Dopamine Receptor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.