missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Pehanon™ pH Paper

Product Code. 11366464 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
1 to 12
1 to 2.8
10.5 to 13
2.8 to 4.6
3.8 to 5.5
4 to 9
5.2 to 6.8
6 to 8.1
7.2 to 8.8
8 to 9.7
9.5 to 12
Unit Size:
Pack of 200
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11366464 3.8 to 5.5 Pack of 200
10004760 10.5 to 13 Pack of 200
10196160 5.2 to 6.8 Pack of 200
10547502 4 to 9 Pack of 200
10708911 6 to 8.1 Pack of 200
10058100 2.8 to 4.6 Pack of 200
10736991 8 to 9.7 Pack of 200
11817043 1 to 12 Pack of 200
10460431 1 to 2.8 Pack of 200
10741601 7.2 to 8.8 Pack of 200
10687901 9.5 to 12 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11366464 missing translation for 'mfr' Macherey-Nagel™ missing translation for 'supplierNo' 90415

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Measure pH in colored solutions. Pehanon™ pH paper puts the indicator and reference on one strip making it perfect for pH determinations in colored solutions.

PEHANON™ is a special pH-test strips that unifies the pH-indicator and the reference colors on one strip and has the following advantages:

Any sample color has the same effect on both, the reference colors and the reactive pad. This ensures unadulterated readings in colored solutions. The user can be sure to get accurate pH-values.

An invisible hydrophobic barrier just above the top color field prevents capillary action of the test solution beyond. The handle will remain dry and clean whatever the sample is. Therefore the use of the strip is very safe.

pH-values can be determined without a separate color chart. Workers and machinists can use single strips instead of complete packs with color chart which makes PEHANON™ highly economic.

Spécification

pH Range 3.8 to 5.5
Format Stick
For Use With (Application) Determination of pH in the colored solutions
Graduations 3.8, 4.0, 4.2, 4.4, 4.6, 4.9, 5.2, 5.5
Quantity 1 Each
Type pH Paper
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.