missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PEI STAR-Go™ transfection reagent

Product Code. 30058548
Change view
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30058548 5 mL 5mL
30058551 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30058548 Supplier R&D Systems™ Supplier No. 8174/5ML
  • ONLINE EXCLUSIVE PRICE
    305.91 €
    266.70 € / 5mL
    Save 39.21 €
    12% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Aqueous solution of polyethylenimine (PEI) transfection reagent

PEI STAR-Go™ is a ready-to-use aqueous formulation of our high performance PEI STAR™ transfection reagent (Cat. No. 7854), provided as a sterile-filtered solution. PEI STAR™ is a chemically-defined, linear polyethylenimine (PEI) transfection reagent for affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293, CHO, and a range of other eukaryotic cell lines. Applications include production of lentivirus and rAAV, recombinant proteins, and antibodies. Supplied as a 1mg/mL aqueous solution

PEI STAR-Go is a trademark of Bio-Techne Corp.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Poly[imino(1,2-ethanediyl)] hydrochloride
CAS 49553-93-7
Quantity 5 mL
Product Line Tocris
Target Transfection Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.