missing translation for 'onlineSavingsMsg'
Learn More

Peimine, MedChemExpress

Product Code. 16431338
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431338 5 mg 5mg
16428898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431338 Supplier Medchem Express Supplier No. HYN0212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Peimine(Dihydroisoimperialine; Verticine) is a natural compound with good anti-inflammatory effects in vivo.

Peimine(Dihydroisoimperialine

Specifications

Chemical Name or Material Peimine
CAS 23496-41-5
Color White
Purity Grade Notes Research
Molecular Formula C27H45NO3
Quantity 5 mg
Synonym Verticine Dihydroisoimperialine
Solubility Information DMSO : 50 mg/mL (115.83 mM; Need ultrasonic)
SMILES [H][C@]12[C@]3([H])C[C@H](O)[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])C[C@]([H])1[C@]5([H])CN6C[C@@H](C)CC[C@]([H])6[C@@](C)(O)[C@]([H])5CC2
Molecular Weight (g/mol) 431.65
Formula Weight 431.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.