missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pelitinib
GREENER_CHOICE

Product Code. 18759066
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18759066 25 mg 25mg
18787856 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18759066 Supplier Tocris Bioscience™ Supplier No. 6349/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and irreversible EGFR inhibitor (IC50 = 20-80 nM); orally bioavailable

Pelitinib is a potent irreversible EGFR inhibitor (IC50 = 20-80 nM). Selective for EGFR over Src, MEK/ERK, erbB2, Raf, c-Met and Cdk4 (IC50 values are 282, 800, 1255, 3353, 4100 and >20,000). Inhibits the growth of tumors overexpressing EGFR in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide
CAS 257933-82-7
Synonym EKB-569
Quantity 25 mg
Molecular Formula C24H23ClFN5O2
Formula Weight 467.92
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.