missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pelitinib

Description
Pelitinib is a potent irreversible EGFR inhibitor (IC50 = 20-80 nM). Selective for EGFR over Src, MEK/ERK, erbB2, Raf, c-Met and Cdk4 (IC50 values are 282, 800, 1255, 3353, 4100 and >20,000). Inhibits the growth of tumors overexpressing EGFR in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide |
| CAS | 257933-82-7 |
| Synonym | EKB-569 |
| Quantity | 5 mg |
| Molecular Formula | C24H23ClFN5O2 |
| Formula Weight | 467.92 |
| Purity | 97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?