missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ FLX2 Cable for CH-3 Headset With External PTT Button

Product Code. 17861041 Shop All 3M Products
Change view
Click to view available options
Model:
FLX2-200
Product Type:
FLX2 Cable J11
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Product Type Unit Size
17861041 FLX2-200 FLX2 Cable J11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17861041 Supplier 3M™ Supplier No. 7100185799
  • ONLINE EXCLUSIVE PRICE
    107.00 €
    80.93 € / Pack of 1
    Save 26.07 €
    24% Off
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PELTOR™ FLX2 platform allows workers to pair protective communication CH-3 two-way headsets with an external push-to-talk button.

  • For use with CH-3 Headset with external PTT Button
  • J11 Plug Type
  • Coiled polyurethane cable
  • Aramid fiber reinforced
  • Robust and highly flexible

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) For use with CH-3 Headsets with external PTT Button
For Use With (Application) Enables connection to two-way communication for CH-3 Headsets
Model FLX2-200
Pairs per Package 1 Ea.
Product Type FLX2 Cable J11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.