Learn More
3M™ PELTOR™ Helmet Mounted Earmuff
Product Code. 30305885 Shop All 3M Products
Change view
Click to view available options
Model:
X2P3E
Unit Size:
Each
Product Code. Model Unit Size
30305885 X2P3E Each
1 options
Product Code. 30305885 Supplier 3M™ Supplier No. 7100326939
  • ONLINE EXCLUSIVE PRICE
    34.45 €
    29.30 € / Pack of 1
    Save 5.15 €
    14% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PELTOR™ X2 Earmuffs offer high-performance hearing protection in a slim, modern design. Available in headband, helmet-mounted, and neckband styles to suit user preference and application needs.
  • Provides reliable hearing protection in environments with moderate noise levels.
  • Colour-coded yellow for easy identification within the PELTOR™ X Series.
  • Electrically insulated twin headband design reduces heat build-up and ensures comfort.
  • Headband features durable stainless-steel wire coated in plastic for strength and comfort.
  • Flat, wide headband with soft plastic coating delivers stable, balanced pressure on the head.
  • Dielectric construction with no exposed metal offers protection in electrically sensitive environments.
  • Smooth-sliding adjustment points for easy fitting and repositioning.
  • Wide, soft foam cushions enhance user comfort during extended wear.
  • Slim cup design improves compatibility with other PPE, including hard hats and safety glasses.
  • Helmet-mounted versions available as dielectric (P5) or with metal arms (P3) depending on electrical insulation requirements.
  • Replaceable cushions and foam inserts (Hygiene Kits) help extend product life and maintain hygiene.
TRUSTED_SUSTAINABILITY
Specifications
Model X2P3E
Noise Reduction Rating (NRR) 31 dB
Corded No
Product Type Helmet-mounted Ear Muff
Disposable Reusable
Metal Detectable Yes
Headband Color Orange
Dielectric No
Style Over-the-head
Foldable Yes