missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ Optime™ II Earmuffs

Product Code. 15719697
Change view
Click to view available options
Model:
H520A-472-GB
H520F-409-GQ
H520F-460-GB
H520P3A-410-GQ
H520P3AF-410-GQ
H520P3B-410-GQ
H520P3E-410-GQ-01
H520P3E-467-GB
H520P3EA-410-GQ
H520P3F-410-GQ
H520P3G-410-GQ
H520P3H-410-GQ
H520P3H-467-GB
H520P3K-410-GQ
Unit Size:
Pack of 10
Pack of 20
Pack of 6
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15719697 H520A-472-GB Pack of 10
15710028 H520P3G-410-GQ Pack of 20
15719917 H520P3A-410-GQ Pack of 20
15729697 H520F-409-GQ Pack of 10
15729917 H520P3B-410-GQ Pack of 20
15739697 H520F-460-GB Pack of 10
15739857 H520P3H-467-GB Pack of 10
15739917 H520P3E-410-GQ-01 Pack of 20
15749697 H520P3E-467-GB Pack of 10
15749917 H520P3EA-410-GQ Pack of 20
15759917 H520P3F-410-GQ Pack of 20
15769917 H520P3H-410-GQ Pack of 20
15779917 H520P3K-410-GQ Pack of 20
15799727 H520P3AF-410-GQ Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 15719697 Supplier 3M™ Supplier No. 7000038207
  • ONLINE EXCLUSIVE PRICE
    421.00 €
    258.03 € / Pack of 10
    Save 162.97 €
    38% Off
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for environments with substantial industrial noise or construction machinery, like airports and agricultural work. 3M™ PELTOR™ Optime™ II Earmuffs have been developed for demanding noise-hazard environments.

  • Ideal hearing protection against noise arising from environments with substantial industrial noise or construction machinery, like airports and agricultural work
  • Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy to replace cushions and inserts helps keep them hygienically clean
TRUSTED_SUSTAINABILITY

Specifications

Color Hi-Viz
Certifications/Compliance CE Approved, EN 352-3: 2002
Model H520A-472-GB
Noise Reduction Rating (NRR) 31 dB
Product Type Ear Muffs
Dielectric No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.