missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ Optime™ III Ear Muffs

Product Code. 15229593
Change view
Click to view available options
Color:
Black, Red
Green, Black
Style:
Headband
Helmet-mounted
Neckband
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Style unitSize
15229593 Black, Red Neckband Each
10185044 Black, Red Headband Each
12990295 Green, Black Helmet-mounted Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15229593 Supplier 3M™ Supplier No. 7000039623

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Developed for use in extremely high noise environments, particularly those dominated by high frequency sounds. 3M™ PELTOR™ Optime™ III Ear Muffs are based on technology featuring double cup design to help minimize resonance effect, resulting in excellent high frequency attenuation.

  • Modern, stylish cup design
  • Good speech intelligibility
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts help keep them hygienically clean
  • Unique low-profile headband design helps maintain constant pressure
  • Soft, wide cusions help reduce pressure around the ears and improve comfort and wearability
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Red
Certifications/Compliance CE Approved
For Use With (Application) Mining, Marine, Airports, and Power Station
Plug Shape Oval
Product Type Hearing Protection Optime III
Dielectric Yes
Style Neckband
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.