missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ SportTac™ Replacement Shells

Product Code. 15709937
Change view
Click to view available options
Color:
Black
Red
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15709937 Red Each
15719937 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15709937 Supplier 3M™ Supplier No. 7000107843
  • ONLINE EXCLUSIVE PRICE
    32.00 €
    28.98 € / Pack of 1
    Save 3.02 €
    9% Off
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

3M™ PELTOR™ SportTac™ Replacement Shells are interchangeable shells that makes it easy to change the color of earmuffs.

  • Clip on replacement shells for 3M™ PELTOR™ SportTac™ Hearing Protectors
  • Interchangeable shells makes it easy to change color
  • One pair per pack
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Product Type Shells
For Use With (Equipment) PELTOR™ SportTac™ Hearing Protectors
Includes One Pair
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.