missing translation for 'onlineSavingsMsg'
Learn More

Pempidine, MedChemExpress

Product Code. 16448746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448746 Supplier Medchem Express Supplier No. HYB1382
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pempidine is a ganglion-blocking drug, introduced as an oral treatment for hypertension.

Pempidine is a ganglion-blocking drug, introduced as an oral treatment for hypertension.

Specifications

Chemical Name or Material Pempidine
CAS 79-55-0
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H21N
Quantity 100 mg
Synonym 1,2,2,6,6-Pentamethylpiperidine
Solubility Information DMSO : ≥ 2.1 mg/mL (13.52 mM)
SMILES CC1(C)CCCC(C)(C)N1C
Molecular Weight (g/mol) 155.28
Formula Weight 155.28
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.