Learn More
Sampling Systems™ Pendulum Beaker
Product Code. 17922042 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Graduated:
No
Yes
Material:
Polypropylene
Stainless Steel
Unit Size:
Each
Product Code. 17922042
Brand: Sampling Systems™ 5065C1001B
Only null left
Add to basket
Add to basket
These pendulum beakers automatically swing to the horizontal position, meaning that your water samples are less likely to spill when using the TeleScoop.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Product Type Swabbing Template
Material Stainless Steel
Autoclavable Autoclavable
Capacity (Metric) 1 L
Disposable Reusable
Graduated No
Sterility Non-sterile