Learn More
Sampling Systems™ Pendulum Beaker
Product Code. 17932042 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Graduated:
No
Yes
Material:
Polypropylene
Stainless Steel
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduated Material Unit Size
17932042 250 mL Yes Polypropylene Each
17902122 500 mL No Polypropylene Each
17922042 1 L No Stainless Steel Each
17942042 1 L No Polypropylene Each
17972042 2 L No Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17932042 Supplier Sampling Systems™ Supplier No. 5065C252
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These pendulum beakers automatically swing to the horizontal position, meaning that your water samples are less likely to spill when using the TeleScoop.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Product Type Swabbing Template
Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 250 mL
Disposable Reusable
Graduated Yes
Sterility Non-sterile