missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V, TRC

Product Code. 30132623
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132623 1 g 1g
30011427 250 mg 250mg
30011849 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132623 Supplier Toronto Research Chemicals Supplier No. TRCP2235051G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-08-1
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-, (2S,5R,6R)-∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)- (8CI)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenoxyacetyl)amino]-, (2S,5R,6R)- (9CI)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenoxyacetyl)amino]-, [2S-(2α,5α,6β)]-∣(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid∣6-(Phenoxyacetamido)penicillanic acid∣Acipen-V∣Calcipen∣Crystapen V∣Distaquaine V∣Eskacillin V∣Fenacilin∣Meropenin∣Oracillin∣Oracilline∣Oratren∣Orocillin∣Ospen∣Pen-Oral∣Pen-Vee∣Pen-vee-oral∣Penicillin V∣Penicillin, phenoxymethyl-∣Phenocillin∣Phenopenicillin∣Phenospen∣Phenoxymethylpenicillin∣Phenoxymethylpenicillinic acid∣Stabicillin∣V-Cil∣V-Cillin∣V-Cylina∣V-Cyline∣V-Tablopen∣Vebecillin∣(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Formula C16H17KN2O5S
Molecular Weight (g/mol) 350.39, 388.48
InChI Key HCTVWSOKIJULET-LQDWTQKMSA-M
SMILES [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
IUPAC Name potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Product Type Metabolite∣API
Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.