missing translation for 'onlineSavingsMsg'
Learn More

Pentacene, TRC

Product Code. 30413117
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413117 Supplier Toronto Research Chemicals Supplier No. TRCP2377705G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentacene
CAS 135-48-8
Molecular Formula C22H14
Quantity 5 g
Synonym 2,3:6,7-Dibenzanthracene, Benzo[b]naphthacene, Dibenz[b,i]anthracene, NSC 90784, lin-Dibenzanthracene, lin-Naphthoanthracene
InChI Formula InChI=1S/C22H14/c1-2-6-16-10-20-14-22-12-18-8-4-3-7-17(18)11-21(22)13-19(20)9-15(16)5-1/h1-14H
SMILES c1ccc2cc3cc4cc5ccccc5cc4cc3cc2c1
IUPAC Name pentacene
Molecular Weight (g/mol) 278.35
Formula Weight 278.1096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.