missing translation for 'onlineSavingsMsg'
Learn More

Pentachloronitrobenzene, 98%, MP Biomedicals

Product Code. 11488425 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488425 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488425 Supplier MP Biomedicals Supplier No. 0522090780
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 82-68-8
Molecular Formula C6Cl5NO2
Quantity 100 g
Synonym pentachloronitrobenzene, quintozene, pcnb, brassicol, terrachlor, batrilex, botrilex, earthcide, fungiclor, pentagen
InChI Key LKPLKUMXSAEKID-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]
IUPAC Name 1,2,3,4,5-pentachloro-6-nitrobenzene
Molecular Weight (g/mol) 295.321
PubChem CID 6720
ChEBI CHEBI:34908
Formula Weight 295.34
Chemical Name or Material Pentachloronitrobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • PENTACHLORONITROBENZENE
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.