missing translation for 'onlineSavingsMsg'
Learn More

Pentachlorophenol, TRC

Product Code. 30411604
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411604 1 g -
30425642 100 mg 100mg
30411502 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411604 Supplier Toronto Research Chemicals Supplier No. TRCP2381001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-86-5
Molecular Formula C6 H Cl5 O
Molecular Weight (g/mol) 266.34
Synonym Phenol, 2,3,4,5,6-pentachloro-, Phenol, pentachloro- (8CI,9CI), 2,3,4,5,6-Pentachlorophenol, 1-Hydroxy-2,3,4,5,6-pentachlorobenzene, 1-Hydroxypentachlorobenzene, Chlon, Chlorophenasic acid, Dowicide 7, Dowicide EC 7, Dura Treet II, EP 30, EP 30 (pesticide), Fungifen, Grundier Arbezol, Lauxtol, Liroprem, NSC 263497, PCP, PCP (pesticide), PKhF, Penchlorol, Penta, Pentachlorophenol, Penton 70, Pentor 70, Perchlorophenol, Permasan, Pol Nu, Pole topper, Pole topper fluid, Preventol P, Santophen 20, Satophen, Witophen P, Woodtreat A
IUPAC Name 2,3,4,5,6-pentachlorophenol
SMILES Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
Formula Weight 263.847
Percent Purity >95%
Chemical Name or Material Pentachlorophenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.