missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanal, TRC

Product Code. 30416434
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416434 250 mg 250mg
30409850 1 g 1g
30419235 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416434 Supplier Toronto Research Chemicals Supplier No. TRCP227100250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2765-11-9
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.4
Synonym 1-Pentadecanal, Pentadecaldehyde, n-Pentadecanal
IUPAC Name pentadecanal
SMILES CCCCCCCCCCCCCCC=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h15H,2-14H2,1H3
Formula Weight 226.2297
Chemical Name or Material Pentadecanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.