missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanoic acid, MedChemExpress

Product Code. 16443419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443419 Supplier Medchem Express Supplier No. HYW004283
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone.

Pentadecylic acid is a saturated fatty acid with a 15-carbon backbone.

Specifications

Chemical Name or Material Pentadecanoic acid
CAS 1002-84-2
Color White
Purity Grade Notes Research
Molecular Formula C15H30O2
Quantity 1 g
Solubility Information DMSO : 62.5 mg/mL (257.84 mM; Need ultrasonic)
SMILES O=C(CCCCCCCCCCCCCC)O
Molecular Weight (g/mol) 242.4
Formula Weight 242.4
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.