missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanoic Acid, TRC

Product Code. 30418816
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418816 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418816 Supplier Toronto Research Chemicals Supplier No. TRCP22048010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentadecanoic acid (Pentadecylic acid)
CAS 1002-84-2
Molecular Formula C15 H30 O2
Quantity 10 g
Synonym Pentadecanoic acid (8CI','9CI','ACI), 14FA, n-Pentadecanoic acid, NSC 28486, Pentadecylic acid
InChI Formula InChI=1S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)
SMILES CCCCCCCCCCCCCCC(=O)O
IUPAC Name pentadecanoic acid
Molecular Weight (g/mol) 242.4
Formula Weight 242.2246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.