missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol, 98+%

Product Code. 11448783
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448783 250 g 250g
11438783 1000 g 1000g
11458783 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448783 Supplier Thermo Scientific Alfa Aesar Supplier No. A10520.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 115-77-5 | C5H12O4 | 136.147 g/mol

Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

Solubility
Soluble in Water: 0.1 g/mL

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00004692
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Spezifikation

Melting Point 260°C to 264°C
Density 1.396
Boiling Point 276°C (30 mmHg)
Flash Point 240°C (464°F)
Quantity 250 g
Beilstein 1679274
Sensitivity Hygroscopic
Merck Index 14,7111
Solubility Information Soluble in Water: 0.1g/mL
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material Pentaerythritol
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.