missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol, 98%

Product Code. 10051300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051300 1 kg Plastic bottle 1kg
10234940 250 g Plastic bottle 250g
11984631 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051300 Supplier Thermo Scientific Acros Supplier No. 129870010
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115-77-5 | C5H12O4 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.15
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Specifications

Melting Point 257.0°C to 264.0°C
Color White
Boiling Point 276.0°C (30.0 mmHg)
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C(CH2OH)4
Quantity 1 kg
Beilstein 01,528
Merck Index 15,7221
Solubility Information Solubility in water: 1 g/18 mL water (15°C). Other solubilities: slightly soluble in alcohol and formamide, insoluble in benzene, carbon tetrachloride, ether,, acetone and petroleum ether
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Pentaerythritol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.