missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol, TRC

Product Code. 30427959
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427959 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427959 Supplier Toronto Research Chemicals Supplier No. TRCP227200250G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentaerythritol
CAS 115-77-5
Molecular Formula C5 H12 O4
Quantity 250 g
Synonym Pentaerythritol (8CI), 2,2-Bis(hydroxymethyl)-1,3-propanediol, Auxinutril, CP 500G, Charmor PM, Charmor PM 15, Charmor PM 40, Flammex DPE, Hercules P 6, Maxinutril, Metab-Auxil, Microlon 93, Monopentaerythritol, Monopentek, NSC 8100, Neulizer P, P 100, P 100 (alcohol), PE 200, PE 200 (diol), PER, PET 020, Penetek, Penta Powder 325, Penta-R, Pentarit, Pentarit S, Pentarit T, Pentek, THME, Tetra(hydroxymethyl)methane, Tetrakis(hydroxymethyl)methane, Tetramethylolmethane, Voxtar M 100
InChI Formula InChI=1S/C5H12O4/c6-1-5(2-7,3-8)4-9/h6-9H,1-4H2
SMILES OCC(CO)(CO)CO
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 136.1464
Formula Weight 136.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.