Learn More
Pentaerythritol triacrylate, stab. with 4-methoxyphenol
Beschreibung
Pentaerythritol triacrylate is used in UV-cured inks, dental polymers and varnishes. Further, it is used in the preparation of printing inks, coatings, in photoresists, acrylic glues, adhesives, binders, paper and wood impregnates. In addition, it serves as a modifier for polyester and fiberglass and plastic components for the medical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| CAS | 3524-68-3 |
| Color | Colorless |
| Density | 1.16 g/mL |
| Flash Point | >110°C (230°F) |
| Molecular Formula | C14H18O7 |
| Refractive Index | 1.484 |
| MDL Number | MFCD00009607 |
| Quantity | 500 g |
| Beilstein | 6598752 |
| Sensitivity | Hygroscopic |
| Mehr anzeigen |
Sicherheit und Handhabung
- Pentaerythritol triacrylate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Pentaerythritol tetraacrylate, Pentaerythritol triacrylate,Trimethylolpropane triacrylate
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.