missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythrityl tetrachloride, 97%, Thermo Scientific™

Product Code. 11427837
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427837 25 g 25g
11417837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427837 Supplier Thermo Scientific Alfa Aesar Supplier No. A16514.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Neopentyl bromide and pentaerythrityl tetrachloride were shown here to be non-mutagenic to 7 strains of Salmonella typhimurium.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3228-99-7
Molecular Formula C5H8Cl4
Molecular Weight (g/mol) 209.92
MDL Number MFCD00000942
InChI Key KPZGRMZPZLOPBS-UHFFFAOYSA-N
Synonym pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane
PubChem CID 76700
IUPAC Name 1,3-dichloro-2,2-bis(chloromethyl)propane
SMILES ClCC(CCl)(CCl)CCl

Specifications

Melting Point 94°C to 96°C
Boiling Point 110°C (12 mmHg)
Quantity 25 g
Beilstein 1698943
Solubility Information Insoluble in water.
Formula Weight 209.93
Percent Purity 97%
Chemical Name or Material Pentaerythrityl tetrachloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.