missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene glycol di-p-toluenesulfonate, 90+%

Product Code. 11323506
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
This item is not returnable. View return policy

Product Code. 11323506

missing translation for 'mfr': Thermo Scientific Alfa Aesar L14369.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41024-91-3 | C24H34O10S2 | 546.646 g/mol

Pentaethylene glycol di-p-toluenesulfonate is used in the preparation of calix[4]arenes, calix[5]arenes, and crown ethers. It is also used in the preparation of 3?-formylbenzo-18-crown-6 by undergoing coupling reaction with 2,3-dihydroxybenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaethylene glycol di-p-toluenesulfonate is used in the preparation of calix[4]arenes, calix[5]arenes, and crown ethers. It is also used in the preparation of 3′-formylbenzo-18-crown-6 by undergoing coupling reaction with 2,3-dihydroxybenzaldehyde.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41024-91-3
Molecular Formula C24H34O10S2
Molecular Weight (g/mol) 546.646
MDL Number MFCD00012204
InChI Key BUHGDYPBQWWWQS-UHFFFAOYSA-N
Synonym 3,6,9,12-tetraoxatetradecane-1,14-diyl bis 4-methylbenzenesulfonate, pentaethylene glycol ditosylate, pentaethylene glycol di-p-toluenesulfonate, pentaethylene glycol di p-toluenesulfonate, tos-peg6-tos, 3,6,9,12-tetraoxatetradecane-1,14-diol, bis 4-methylbenzenesulfonate, pentaethylene glycol d p-toluenesulfonate, penta ethylene glycol di-p-toluenesulfonate, 3,6,9,12-tetraoxatetradecane-1,14-diyl ditosylate
PubChem CID 361968
IUPAC Name 2-[2-[2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Density 1.26
Flash Point >110°C (230°F)
Refractive Index 1.5225
Quantity 5 g
Beilstein 3599245
Sensitivity Hygroscopic
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 546.66
Percent Purity 95%
Chemical Name or Material Pentaethylene glycol di-p-toluenesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.