missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzenesulfonyl chloride, 99%

Product Code. 10603892 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10603892 25 g Glass bottle 25g
10428872 1 g Glass bottle 1g
10503311 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10603892 Supplier Thermo Scientific Supplier No. 147110250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 832-53-1 | C6ClF5O2S | 266.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832-53-1
Molecular Formula C6ClF5O2S
Molecular Weight (g/mol) 266.57
MDL Number MFCD00007427
InChI Key UOJCTEGNHXRPKO-UHFFFAOYSA-N
Synonym pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
PubChem CID 70026
IUPAC Name 2,3,4,5,6-pentafluorobenzenesulfonyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F

Specifications

Density 1.7960g/mL
Boiling Point 211°C
Flash Point 90°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6F5SO2Cl
Refractive Index 1.4780 to 1.48
Quantity 25 g
Specific Gravity 1.796
Formula Weight 266.57
Percent Purity 99%
Chemical Name or Material Pentafluorobenzenesulfonyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Pentafluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.