missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl Chloride, TRC

Product Code. 30414638
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414638 1 g 1g
30409612 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414638 Supplier Toronto Research Chemicals Supplier No. TRCP2703001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251-50-5
Molecular Formula C7 Cl F5 O
Molecular Weight (g/mol) 230.52
Synonym 2,3,4,5,6-Pentafluorobenzoic acid chloride, 2,3,4,5,6-Pentafluorobenzoyl chloride, NSC 97002, Perfluorobenzoyl Chloride
IUPAC Name 2,3,4,5,6-pentafluorobenzoyl chloride
SMILES Fc1c(F)c(F)c(C(=O)Cl)c(F)c1F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7ClF5O/c8-7(14)1-2(9)4(11)6(13)5(12)3(1)10
Formula Weight 229.9558
Chemical Name or Material Pentafluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.