missing translation for 'onlineSavingsMsg'
Learn More

(Pentafluoroethyl)trimethylsilane, 97%

Product Code. 11454350
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454350 5 g 5g
11444350 1 g 1g
11464350 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454350 Supplier Thermo Scientific Alfa Aesar Supplier No. H25775.06
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124898-13-1 | C5H9F5Si | 192.204 g/mol

(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.

Solubility
Miscible with tetrahydrofuran.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124898-13-1
Molecular Formula C5H9F5Si
Molecular Weight (g/mol) 192.204
MDL Number MFCD04116436
InChI Key MTPVUVINMAGMJL-UHFFFAOYSA-N
Synonym pentafluoroethyl trimethylsilane, trimethyl pentafluoroethyl silane, pentafluoroethyltrimethylsilane, trimethylpentafluoroethylsilane, trimethyl perfluoroethyl silane, trimethyl 1,1,2,2,2-pentafluoroethyl silane, silane, trimethyl pentafluoroethyl, silane,trimethyl 1,1,2,2,2-pentafluoroethyl, acmc-20aple, trimethylsilylpentafluoroethane
PubChem CID 2760820
IUPAC Name trimethyl(1,1,2,2,2-pentafluoroethyl)silane
SMILES C[Si](C)(C)C(C(F)(F)F)(F)F

Specifications

Quantity 5 g
Refractive Index 1.327
Sensitivity Moisture sensitive
UN Number UN1993
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material (Pentafluoroethyl)trimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.