missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate, 90%, Thermo Scientific™

Product Code. 11336835
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11336835 5 g Amber Glass Bottle 5g
10215574 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336835 Supplier Thermo Scientific Maybridge Supplier No. CC50626DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-81-1
Molecular Formula C21H20F5N3O4
Molecular Weight (g/mol) 473.4
MDL Number MFCD09966147
InChI Key XHATYMDMSNXMKQ-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-4-tert-butoxycarbonyl piperazin-1-yl isonicotinate, tert-butyl 4-4-pentafluorophenoxycarbonyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-pentafluorophenoxy carbonyl pyridin-2-yl piperazine-1-carboxylate, 4-4-oxo-2,3,4,5,6-pentafluorophenoxy methyl-2-pyridinyl-1-piperazinecarboxylic acid tert-butyl ester, tert-butyl 4-4-2,3,4,5,6-pentafluorophenoxycarbonyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-2,3,4,5,6-pentakis fluoranyl phenoxy carbonylpyridin-2-yl piperazine-1-carboxylate
PubChem CID 43811038
IUPAC Name tert-butyl 4-[4-(2,3,4,5,6-pentafluorophenoxy)carbonylpyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=CC(=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 473.4
Percent Purity 90%
Chemical Name or Material Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.