missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic anhydride, 98%

Product Code. 11461798
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461798 100 g 100g
11441798 5 g 5g
11451798 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Rückgaberichtlinie anzeigen
Artikelnummer. 11461798 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A14472.22
 Restricted Item
Estimated Shipment: 30-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 356-42-3 | C6F10O3 | 310.047 g/mol

Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

Solubility
Reacts with water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 356-42-3
Molecular Formula C6F10O3
Molecular Weight (g/mol) 310.047
MDL Number MFCD00000429
InChI Key XETRHNFRKCNWAJ-UHFFFAOYSA-N
Synonym pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
PubChem CID 67742
IUPAC Name 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
SMILES C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F

Spezifikation

Density 1.589
Boiling Point 73°C to 74°C
Quantity 100 g
UN Number UN3265
Beilstein 1806446
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 310.04
Percent Purity 98%
Chemical Name or Material Pentafluoropropionic anhydride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.