missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic anhydride, 98%

Product Code. 10521892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521892 5 g Glass bottle 5g
10529292 25 g Glass bottle 25g
11964941 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10521892 Supplier Thermo Scientific Acros Supplier No. 416930050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 356-42-3 | C6F10O3 | 310.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356-42-3
Molecular Formula C6F10O3
Molecular Weight (g/mol) 310.04
InChI Key XETRHNFRKCNWAJ-UHFFFAOYSA-N
Synonym pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride
PubChem CID 67742
IUPAC Name 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate
SMILES C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F

Specifications

Color Colorless
Density 1.5700g/mL
Boiling Point 69.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CF3CF2CO)2O
Refractive Index 1.3
Quantity 5 g
Specific Gravity 1.57
Formula Weight 310.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentafluoropropionic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.