missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropyridine, 99%

Product Code. 11484197
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484197 5 g 5g
11494197 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484197 Supplier Thermo Scientific Alfa Aesar Supplier No. A19656.06
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 700-16-3 | C5F5N | 169.054 g/mol

Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, alkali metals and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-16-3
Molecular Formula C5F5N
Molecular Weight (g/mol) 169.054
MDL Number MFCD00006225
InChI Key XTGOWLIKIQLYRG-UHFFFAOYSA-N
Synonym pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
PubChem CID 69690
IUPAC Name 2,3,4,5,6-pentafluoropyridine
SMILES C1(=C(C(=NC(=C1F)F)F)F)F

Specifications

Density 1.618
Boiling Point 83°C to 85°C
Flash Point 23°C (73°F)
Refractive Index 1.3865
Quantity 5 g
UN Number UN2924
Beilstein 1427064
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 169.05
Percent Purity 99%
Chemical Name or Material Pentafluoropyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentafluoropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Pentafluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.