missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropyridine, 99+%

Product Code. 10774353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10774353 25 g Glass bottle 25g
10295500 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10774353 Supplier Thermo Scientific Acros Supplier No. 169990250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 700-16-3 | C5F5N | 169.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-16-3
Molecular Formula C5F5N
Molecular Weight (g/mol) 169.05
MDL Number MFCD00006225
InChI Key XTGOWLIKIQLYRG-UHFFFAOYSA-N
Synonym pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
PubChem CID 69690
IUPAC Name 2,3,4,5,6-pentafluoropyridine
SMILES C1(=C(C(=NC(=C1F)F)F)F)F

Specifications

Color Colorless
Density 1.5400g/mL
Boiling Point 84°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (C5N)F5
Refractive Index 1.3850 to 1.387
Quantity 25 g
Specific Gravity 1.54
Formula Weight 169.05
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Pentafluoropyridine, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentafluoropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.